missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzo[d]isoxazol-3-ol, 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC450960050

 View more versions of this product

Catalog No. AC450960050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Benzo[d]isoxazol-3-ol
5g
95% min. (HPLC)
1,2-benzisoxazol-3 2h-one, 1,2-benzisoxazol-3-ol, 1,2-benzisoxazol-3-ol 8ci, 1,2-benzisoxazolin-3-one, 1,2-benzoxazol-3-ol, 2h-1,2-benzoxazol-3-one, 3-hydroxy-1,2-benzisoxazole, 3-hydroxybenzisoxazole, benzisoxazolone, benzo d isoxazol-3-ol
C1=CC=C2C(=C1)C(=O)NO2
135.12
135.12
137°C to 140°C
21725-69-9
C7H5NO2
QLDQYRDCPNBPII-UHFFFAOYSA-N
1,2-benzoxazol-3-one
210830
96%

Chemical Identifiers

21725-69-9
135.12
1,2-benzisoxazol-3 2h-one, 1,2-benzisoxazol-3-ol, 1,2-benzisoxazol-3-ol 8ci, 1,2-benzisoxazolin-3-one, 1,2-benzoxazol-3-ol, 2h-1,2-benzoxazol-3-one, 3-hydroxy-1,2-benzisoxazole, 3-hydroxybenzisoxazole, benzisoxazolone, benzo d isoxazol-3-ol
1,2-benzoxazol-3-one
C7H5NO2
QLDQYRDCPNBPII-UHFFFAOYSA-N
210830
C1=CC=C2C(=C1)C(=O)NO2
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

SDS Documents