Please login to your online account to display your discounted pricing

Benzo[d]isoxazol-3-ol, ≥95%, Maybridge

Chemical Identifiers

Chemical Name or Material Benzo[d]isoxazol-3-ol
CAS 21725-69-9
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C7H5NO2
Formula Weight 135.12
MDL Number MFCD00125030
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RF01278EA

 
fisher scientific company
RF01278EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 135.12
Quantity 10g
Assay 95%
CAS Min % 95.0
Packaging Amber Glass Bottle
Molecular Formula C7H5NO2
MDL Number MFCD00125030
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Chemical Name or Material Benzo[d]isoxazol-3-ol
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 95%
CAS 21725-69-9