Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzo[c]cinnoline N-oxide, 95%

$133.00 - $133.00

Chemical Identifiers

Chemical Name or Material Benzo[c]cinnoline N-oxide
CAS 6141-98-6
Molecular Formula C12H8N2O
Formula Weight 196.21
MDL Number MFCD00014475
Beilstein 8732
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2254706

  SDS
alfa aesar™
B2254706
5g Each for $133.00
Description & Specifications

Specifications

Formula Weight 196.21
Chemical Name or Material Benzo[c]cinnoline N-oxide
CAS 6141-98-6
Beilstein 8732
Melting Point 138° to 141°C
Quantity 5g
RTECS Number DE7644000
Assay 95%
Molecular Formula C12H8N2O
MDL Number MFCD00014475
Odor Odorless