Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzo[c]cinnoline, 99%

$125.51 - $125.51

Chemical Identifiers

Chemical Name or Material Benzo[c]cinnoline
CAS 230-17-1
Molecular Formula C12H8N2
Formula Weight 180.21
MDL Number MFCD00004990
Beilstein 126519
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0040903

  SDS
alfa aesar™
L0040903
1g Each for $125.51
Description & Specifications

Specifications

Formula Weight 180.21
Chemical Name or Material Benzo[c]cinnoline
CAS 230-17-1
MDL Number MFCD00004990
Assay 99%
Molecular Formula C12H8N2
Odor Odorless
Melting Point 156° to 160°C
Beilstein 126519
Quantity 1g