missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzo[b]thiophene-3-sulfonyl chloride, 97%, Alfa Aesar™

$96.69 - $305.34

Chemical Identifiers

CAS 18494-87-6
Molecular Formula C8H5ClO2S2
Molecular Weight (g/mol) 232.696
MDL Number MFCD02681897
InChI Key WYYRQMZTUXFUTC-UHFFFAOYSA-N
Synonym benzothiophene-3-sulfonyl chloride, benzo b thiophene-3-sulfonyl chloride, benzo b thiophene-3-sulfonylchloride, benzo b thiophene-3-sulphonyl chloride, pubchem14540, 3-chlorosulphonyl-benzothiophene, benzothiophene-3-sulfonylchloride, 3-chlorosulphonyl-1-benzothiophene, benzo b thiophen-3-ylchlorosulfone, 3-chlorosulphonyl-1-benzothiophene
PubChem CID 2776340
IUPAC Name 1-benzothiophene-3-sulfonyl chloride
SMILES C1=CC=C2C(=C1)C(=CS2)S(=O)(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH61368MD View Documents Alfa Aesar
H61368MD
250mg Each for $96.69
 
AAH6136803 View Documents Alfa Aesar
H6136803
1g Each for $305.34
 
Description

Description

Applications
Benzo[b]thiophene-3-sulfonyl chloride is used as a pharmaceutical intermediate.

Solubility
Sparingly soluble in water (0.091 g/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture.
Specifications

Chemical Identifiers

18494-87-6
232.696
WYYRQMZTUXFUTC-UHFFFAOYSA-N
2776340
C1=CC=C2C(=C1)C(=CS2)S(=O)(=O)Cl
C8H5ClO2S2
MFCD02681897
benzothiophene-3-sulfonyl chloride, benzo b thiophene-3-sulfonyl chloride, benzo b thiophene-3-sulfonylchloride, benzo b thiophene-3-sulphonyl chloride, pubchem14540, 3-chlorosulphonyl-benzothiophene, benzothiophene-3-sulfonylchloride, 3-chlorosulphonyl-1-benzothiophene, benzo b thiophen-3-ylchlorosulfone, 3-chlorosulphonyl-1-benzothiophene
1-benzothiophene-3-sulfonyl chloride

Specifications

86°C to 88°C
18494-87-6
MFCD02681897
1285265
WYYRQMZTUXFUTC-UHFFFAOYSA-N
1-benzothiophene-3-sulfonyl chloride
2776340
97%
250mg
C8H5ClO2S2
UN3261
benzothiophene-3-sulfonyl chloride, benzo b thiophene-3-sulfonyl chloride, benzo b thiophene-3-sulfonylchloride, benzo b thiophene-3-sulphonyl chloride, pubchem14540, 3-chlorosulphonyl-benzothiophene, benzothiophene-3-sulfonylchloride, 3-chlorosulphonyl-1-benzothiophene, benzo b thiophen-3-ylchlorosulfone, 3-chlorosulphonyl-1-benzothiophene
C1=CC=C2C(=C1)C(=CS2)S(=O)(=O)Cl
232.696
232.71
Benzo[b]thiophene-3-sulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Recommended Storage : 2° to 8°C