missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzo[b]thiophene-3-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6136803

 View more versions of this product

Catalog No. AAH6136803



Description

Description

Benzo[b]thiophene-3-sulfonyl chloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzo[b]thiophene-3-sulfonyl chloride is used as a pharmaceutical intermediate.

Solubility
Sparingly soluble in water (0.091 g/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture.
Specifications

Specifications

Benzo[b]thiophene-3-sulfonyl chloride
86°C to 88°C
MFCD02681897
UN3261
Moisture sensitive
Sparingly soluble in water (0.091g/L at 25°C).
C1=CC=C2C(=C1)C(=CS2)S(=O)(=O)Cl
232.696
232.71
18494-87-6
C8H5ClO2S2
1 g
1285265
benzothiophene-3-sulfonyl chloride, benzo b thiophene-3-sulfonyl chloride, benzo b thiophene-3-sulfonylchloride, benzo b thiophene-3-sulphonyl chloride, pubchem14540, 3-chlorosulphonyl-benzothiophene, benzothiophene-3-sulfonylchloride, 3-chlorosulphonyl-1-benzothiophene, benzo b thiophen-3-ylchlorosulfone
WYYRQMZTUXFUTC-UHFFFAOYSA-N
1-benzothiophene-3-sulfonyl chloride
2776340
97%

Chemical Identifiers

18494-87-6
232.696
WYYRQMZTUXFUTC-UHFFFAOYSA-N
2776340
C1=CC=C2C(=C1)C(=CS2)S(=O)(=O)Cl
C8H5ClO2S2
MFCD02681897
benzothiophene-3-sulfonyl chloride, benzo b thiophene-3-sulfonyl chloride, benzo b thiophene-3-sulfonylchloride, benzo b thiophene-3-sulphonyl chloride, pubchem14540, 3-chlorosulphonyl-benzothiophene, benzothiophene-3-sulfonylchloride, 3-chlorosulphonyl-1-benzothiophene, benzo b thiophen-3-ylchlorosulfone
1-benzothiophene-3-sulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only