Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzo[b]thiophene-3-acetonitrile, 98+%

Chemical Identifiers

Chemical Name or Material Benzo[b]thiophene-3-acetonitrile
CAS 3216-48-6
Molecular Formula C10H7NS
Formula Weight 173.24
MDL Number MFCD00027389
UN Number UN3439
Beilstein 125487
Synonym 3-Cyanomethylthianaphthene; Thianaphthene-3-acetonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0854703

  SDS
alfa aesar™
L0854703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0854706

  SDS
alfa aesar™
L0854706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 173.24
Chemical Name or Material Benzo[b]thiophene-3-acetonitrile
Assay ≥98%
CAS 3216-48-6
Molecular Formula C10H7NS
MDL Number MFCD00027389
UN Number UN3439
Beilstein 125487
Synonym 3-Cyanomethylthianaphthene; Thianaphthene-3-acetonitrile
Melting Point 62° to 66°C
Health Hazard 1 Danger
Quantity 1g
Health Hazard 2 GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Applications
Benzo[b]thiophene-3-acetonitrile is used to produce 2-(5-Bromo-benzo[b]thiophen-3-yl)-ethylamine; hydrochloride. It is also used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Incompatible with oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Safety and Handling

Danger

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.