Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzo[b]thiophene-2-methanol, 97%

$113.46 - $409.84

Chemical Identifiers

Chemical Name or Material Benzo[b]thiophene-2-methanol
CAS 17890-56-1
Molecular Formula C9H8OS
Formula Weight 164.22
MDL Number MFCD02682004
Synonym 2-(Hydroxymethyl)benzo[b]thiophene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2751203

  SDS
alfa aesar™
H2751203
1g Each for $113.46

AAH2751206

  SDS
alfa aesar™
H2751206
5g Each for $409.84
Description & Specifications

Specifications

Formula Weight 164.22
MDL Number MFCD02682004
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym 2-(Hydroxymethyl)benzo[b]thiophene
Health Hazard 1 Warning
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Benzo[b]thiophene-2-methanol
CAS 17890-56-1
Melting Point 98° to 101°C
Molecular Formula C9H8OS
Quantity 1g

Applications
Benzo[b]thiophene-2-methano is an important intermediates and reagents in organic synthesis.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.