Please login to your online account to display your discounted pricing

Benzo[b]thiophene-2-carboxylic hydrazide, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Benzo[b]thiophene-2-carboxylic hydrazide
CAS 175135-07-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H8N2OS
Formula Weight 192.24
MDL Number MFCD00052501
Synonym Thianaphthene-2-carboxylic hydrazide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC368660010

  SDS
thermo fisher scientific
AC368660010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Needles
Formula Weight 192.24
Quantity 1g
Color Off-white to beige
CAS Max % 100.0
Chemical Name or Material Benzo[b]thiophene-2-carboxylic hydrazide
Molecular Formula C9H8N2OS
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay Percent Range 99%
MDL Number MFCD00052501
CAS 175135-07-6
Infrared Spectrum Authentic
Synonym Thianaphthene-2-carboxylic hydrazide
Packaging Glass bottle
CAS Min % 98.5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.