Please login to your online account to display your discounted pricing

Benzo[b]thiophene-2-carboxylic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Benzo[b]thiophene-2-carboxylic acid
Name Note 97%
CAS 6314-28-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9 H6 O2 S
Formula Weight 178.21
MDL Number MFCD00051636
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC330060100

  SDS
thermo fisher scientific
33006-0100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 178.21
Physical Form Crystalline Powder
Quantity 10g
Color Off-white to yellow-beige
Merck Index 15, 9498
Melting Point 238° to 243°C
CAS Min % 96.0
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Name Note 97%
Assay Percent Range 97%
CAS 6314-28-9
Chemical Name or Material Benzo[b]thiophene-2-carboxylic acid
Health Hazard 1 STENCH
MDL Number MFCD00051636
Molecular Formula C9 H6 O2 S
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

STENCH

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.