missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzo[b]thiophene-2-boronic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2283506

 View more versions of this product

Catalog No. AAB2283506



Description

Description

Benzo[b]thiophene-2-boronic acid is used in the synthesis of phosphorescent sensor for quantification of copper(II) ion, UV promoted phenanthridine syntheses, Preparation of CYP11B1 inhibitors for treatment of cortisol dependent diseases, PDE4 inhibitors, Suzuki-Miyaura cross-coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzo[b]thiophene-2-boronic acid is used in the synthesis of phosphorescent sensor for quantification of copper(II) ion, UV promoted phenanthridine syntheses, Preparation of CYP11B1 inhibitors for treatment of cortisol dependent diseases, PDE4 inhibitors, Suzuki-Miyaura cross-coupling reactions.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
Specifications

Specifications

Benzo[b]thiophene-2-boronic acid
∼255°C
MFCD01075674
1637924
Insoluble in water.
OB(O)C1=CC2=CC=CC=C2S1
178.01
178.02
98437-23-1
C8H7BO2S
5 g
2-benzothienylboronic acid, benzo b thiophen-2-ylboronic acid, benzo b thiophene-2-boronic acid, benzothiophene-2-boronic acid, 1-benzothien-2-ylboronic acid, thianaphthene-2-boronic acid, benzo b thiophene-2-ylboronic acid, 2-benzothiopheneboronic acid, benzothiophen-2-ylboronic acid
YNCYPMUJDDXIRH-UHFFFAOYSA-N
1-benzothiophen-2-ylboronic acid
2359
98%

Chemical Identifiers

98437-23-1
178.01
YNCYPMUJDDXIRH-UHFFFAOYSA-N
2359
C8H7BO2S
MFCD01075674
2-benzothienylboronic acid, benzo b thiophen-2-ylboronic acid, benzo b thiophene-2-boronic acid, benzothiophene-2-boronic acid, 1-benzothien-2-ylboronic acid, thianaphthene-2-boronic acid, benzo b thiophene-2-ylboronic acid, 2-benzothiopheneboronic acid, benzothiophen-2-ylboronic acid
OB(O)C1=CC2=CC=CC=C2S1
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only