Please login to your online account to display your discounted pricing

Benzo[b]furan-7-sulfonyl chloride, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC78203DE

 View more versions of this product

Catalog No. CC78203DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 216.64
Quantity 5g
Assay 97%
CAS 1191030-88-2
Chemical Name or Material Benzo[b]furan-7-sulfonyl chloride
Health Hazard 1 GHS Signal Word: Danger
Packaging Amber Glass Bottle
Molecular Formula C8H5ClO3S

Specifications

Chemical Name or Material Benzo[b]furan-7-sulfonyl chloride
Assay 97%
Molecular Formula C8H5ClO3S
Formula Weight 216.64
Accessories

GHS Signal Word: Danger

SDS & Safety Documents