Please login to your online account to display your discounted pricing

Benzo[b]furan-7-carboxylic acid, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC78201DA

 View more versions of this product

Catalog No. CC78201DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 162.14
Quantity 1g
Assay 97%
Chemical Name or Material Benzo[b]furan-7-carboxylic acid
Packaging Amber Glass Bottle
Molecular Formula C9H6O3
CAS 90484-22-3

Specifications

Chemical Name or Material Benzo[b]furan-7-carboxylic acid
Assay 97%
Molecular Formula C9H6O3
Formula Weight 162.14