missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Benzo[b]furan-7-carboxylic acid, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC78201DE
Specifications
Benzo[b]furan-7-carboxylic acid | |
Amber Glass Bottle | |
MFCD10000617 | |
benzofuran-7-carboxylic acid, benzofuran-7-carboxylicacid, 7-benzofuranylcarboxylic acid, benzo b furan-7-carboxylic acid, 7-benzofurancarboxylic acid, 4ahs, 7-benzofurancarboxylicacid, akh | |
OC(=O)C1=C2OC=CC2=CC=C1 | |
162.14 | |
162.14 |
90484-22-3 | |
C9H6O3 | |
5g | |
QMHILIQFOBNARN-UHFFFAOYSA-N | |
1-benzofuran-7-carboxylic acid | |
13307983 | |
97% |
Chemical Identifiers
90484-22-3 | |
162.14 | |
QMHILIQFOBNARN-UHFFFAOYSA-N | |
13307983 | |
OC(=O)C1=C2OC=CC2=CC=C1 |
C9H6O3 | |
MFCD10000617 | |
benzofuran-7-carboxylic acid, benzofuran-7-carboxylicacid, 7-benzofuranylcarboxylic acid, benzo b furan-7-carboxylic acid, 7-benzofurancarboxylic acid, 4ahs, 7-benzofurancarboxylicacid, akh | |
1-benzofuran-7-carboxylic acid |