Please login to your online account to display your discounted pricing

Benzo[b]furan-7-carbaldehyde, Acros Organics


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC78204DA

 View more versions of this product

Catalog No. CC78204DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 146.14
Quantity 1g
Health Hazard 3 GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Assay 97%
CAS Max % 100.0
Chemical Name or Material Benzo[b]furan-7-carbaldehyde
Health Hazard 1 Warning
CAS 95333-14-5
CAS Min % 96.5
MDL Number MFCD08272125
Molecular Formula C9H6O2
Packaging Amber Glass Bottle
Assay Percent Range 97%
Health Hazard 2 GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material Benzo[b]furan-7-carbaldehyde
CAS 95333-14-5
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H6O2
Formula Weight 146.14
MDL Number MFCD08272125
Accessories

Warning

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents