missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Benzo[b]furan-7-carbaldehyde, Thermo Scientific™
Chemical Identifiers
CAS | 95333-14-5 |
---|---|
Molecular Formula | C9H6O2 |
Molecular Weight (g/mol) | 146.145 |
MDL Number | MFCD08272125 |
InChI Key | RGPUSZZTRKTMNA-UHFFFAOYSA-N |
Synonym | benzofuran-7-carbaldehyde, 7-benzofurancarboxaldehyde, 7-formylbenzofuran, 7-benzofurancarboxaldehyde 9ci, benzofuran-7-carboxaldehyde, 7-benzo b furancarbaldehyde, benzo b furan-7-carbaldehyde |
PubChem CID | 11126406 |
IUPAC Name | 1-benzofuran-7-carbaldehyde |
SMILES | C1=CC2=C(C(=C1)C=O)OC=C2 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
CC78204DA
|
Thermo Scientific Chemicals
CC78204DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC78204DE
|
Thermo Scientific Chemicals
CC78204DE |
5g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
95333-14-5 | |
146.145 | |
RGPUSZZTRKTMNA-UHFFFAOYSA-N | |
11126406 | |
C1=CC2=C(C(=C1)C=O)OC=C2 |
C9H6O2 | |
MFCD08272125 | |
benzofuran-7-carbaldehyde, 7-benzofurancarboxaldehyde, 7-formylbenzofuran, 7-benzofurancarboxaldehyde 9ci, benzofuran-7-carboxaldehyde, 7-benzo b furancarbaldehyde, benzo b furan-7-carbaldehyde | |
1-benzofuran-7-carbaldehyde |
Specifications
95333-14-5 | |
100.0 | |
Amber Glass Bottle | |
MFCD08272125 | |
benzofuran-7-carbaldehyde, 7-benzofurancarboxaldehyde, 7-formylbenzofuran, 7-benzofurancarboxaldehyde 9ci, benzofuran-7-carboxaldehyde, 7-benzo b furancarbaldehyde, benzo b furan-7-carbaldehyde | |
C1=CC2=C(C(=C1)C=O)OC=C2 | |
146.145 | |
146.14 | |
Benzo[b]furan-7-carbaldehyde |
96.5 | |
95% | |
C9H6O2 | |
1g | |
RGPUSZZTRKTMNA-UHFFFAOYSA-N | |
1-benzofuran-7-carbaldehyde | |
11126406 | |
95% |
Safety and Handling
GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Warning