missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Benzo[b]furan-7-carbaldehyde, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC78204DE
Specifications
Benzo[b]furan-7-carbaldehyde | |
96.5 | |
95% | |
C9H6O2 | |
5g | |
RGPUSZZTRKTMNA-UHFFFAOYSA-N | |
1-benzofuran-7-carbaldehyde | |
11126406 | |
95% |
95333-14-5 | |
100.0 | |
Amber Glass Bottle | |
MFCD08272125 | |
benzofuran-7-carbaldehyde, 7-benzofurancarboxaldehyde, 7-formylbenzofuran, 7-benzofurancarboxaldehyde 9ci, benzofuran-7-carboxaldehyde, 7-benzo b furancarbaldehyde, benzo b furan-7-carbaldehyde | |
C1=CC2=C(C(=C1)C=O)OC=C2 | |
146.145 | |
146.14 |
Chemical Identifiers
95333-14-5 | |
146.145 | |
RGPUSZZTRKTMNA-UHFFFAOYSA-N | |
11126406 | |
C1=CC2=C(C(=C1)C=O)OC=C2 |
C9H6O2 | |
MFCD08272125 | |
benzofuran-7-carbaldehyde, 7-benzofurancarboxaldehyde, 7-formylbenzofuran, 7-benzofurancarboxaldehyde 9ci, benzofuran-7-carboxaldehyde, 7-benzo b furancarbaldehyde, benzo b furan-7-carbaldehyde | |
1-benzofuran-7-carbaldehyde |
Safety and Handling
GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Warning