Please login to your online account to display your discounted pricing

Benzo[b]furan-3-ylacetic acid, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC40801DA

 View more versions of this product

Catalog No. CC40801DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 176.17
Quantity 1g
Assay 97%
Chemical Name or Material Benzo[b]furan-3-ylacetic acid
Molecular Formula C10H8O3
CAS 64175-51-5
Packaging Amber Glass Bottle
Health Hazard 1 Warning
Assay Percent Range 97%

Specifications

Chemical Name or Material Benzo[b]furan-3-ylacetic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H8O3
Formula Weight 176.17
Accessories

Warning

SDS & Safety Documents