Please login to your online account to display your discounted pricing

Benzo[b]furan-3-carbonyl chloride, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC27502CB

 View more versions of this product

Catalog No. CC27502CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 180.59
Quantity 250mg
Assay 97%
CAS Max % 100.0
CAS 111964-21-7
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
CAS Min % 96.5
Assay Percent Range 97%
MDL Number MFCD11841066
Chemical Name or Material Benzo[b]furan-3-carbonyl chloride
Molecular Formula C9H5ClO2
Packaging Amber Glass Bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material Benzo[b]furan-3-carbonyl chloride
CAS 111964-21-7
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H5ClO2
Formula Weight 180.59
MDL Number MFCD11841066
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

SDS & Safety Documents