Learn More
Benzo[b]furan-2-carboxylic acid, 98+%, Alfa Aesar™
Manufacturer: Alfa Aesar A1557906
Specifications
Benzo[b]furan-2-carboxylic acid | |
192°C to 196°C | |
496-41-3 | |
MFCD00005848 | |
14,2561 | |
OFFSPAZVIVZPHU-UHFFFAOYSA-N | |
1-benzofuran-2-carboxylic acid | |
10331 | |
≥98% |
310°C to 315°C | |
5g | |
C9H6O3 | |
124204 | |
benzofuran-2-carboxylic acid, coumarilic acid, benzo b furan-2-carboxylic acid, 2-benzofurancarboxylic acid, 2-carboxybenzofuran, coumarone-2-carboxylic acid, unii-7z6nmg947l, benzo b furan-2-carboxylic acid, chembl84095, benzofuran-2-carboxylicacid | |
C1=CC=C2C(=C1)C=C(O2)C(=O)O | |
162.144 | |
162.15 |
Chemical Identifiers
496-41-3 | |
162.144 | |
OFFSPAZVIVZPHU-UHFFFAOYSA-N | |
10331 | |
C1=CC=C2C(=C1)C=C(O2)C(=O)O |
C9H6O3 | |
MFCD00005848 | |
benzofuran-2-carboxylic acid, coumarilic acid, benzo b furan-2-carboxylic acid, 2-benzofurancarboxylic acid, 2-carboxybenzofuran, coumarone-2-carboxylic acid, unii-7z6nmg947l, benzo b furan-2-carboxylic acid, chembl84095, benzofuran-2-carboxylicacid | |
1-benzofuran-2-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
RTECSNumber : DF6490000
TSCA : TSCA