missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzo[b]furan-2-carboxylic acid, 98+%, Alfa Aesar™

Manufacturer:  Alfa Aesar A1557914

 View more versions of this product

Catalog No. AAA1557914



Specifications

Specifications

Benzo[b]furan-2-carboxylic acid
192°C to 196°C
496-41-3
MFCD00005848
14,2561
OFFSPAZVIVZPHU-UHFFFAOYSA-N
1-benzofuran-2-carboxylic acid
10331
≥98%
310°C to 315°C
25g
C9H6O3
124204
benzofuran-2-carboxylic acid, coumarilic acid, benzo b furan-2-carboxylic acid, 2-benzofurancarboxylic acid, 2-carboxybenzofuran, coumarone-2-carboxylic acid, unii-7z6nmg947l, benzo b furan-2-carboxylic acid, chembl84095, benzofuran-2-carboxylicacid
C1=CC=C2C(=C1)C=C(O2)C(=O)O
162.144
162.15

Chemical Identifiers

496-41-3
162.144
OFFSPAZVIVZPHU-UHFFFAOYSA-N
10331
C1=CC=C2C(=C1)C=C(O2)C(=O)O
C9H6O3
MFCD00005848
benzofuran-2-carboxylic acid, coumarilic acid, benzo b furan-2-carboxylic acid, 2-benzofurancarboxylic acid, 2-carboxybenzofuran, coumarone-2-carboxylic acid, unii-7z6nmg947l, benzo b furan-2-carboxylic acid, chembl84095, benzofuran-2-carboxylicacid
1-benzofuran-2-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DF6490000

TSCA : TSCA

Documents