Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzo[b]furan-2-carboxylic acid, 98+%

$143.59 - $143.59

Chemical Identifiers

Chemical Name or Material Benzo[b]furan-2-carboxylic acid
CAS 496-41-3
Molecular Formula C9H6O3
Formula Weight 162.15
MDL Number MFCD00005848
Beilstein 124204
Merck Index 14,2561
Synonym Coumarilic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1557906

  SDS
alfa aesar™
A1557906
5g Each for $143.59
Description & Specifications

Specifications

Formula Weight 162.15
Chemical Name or Material Benzo[b]furan-2-carboxylic acid
TSCA TSCA
Assay ≥98%
Molecular Formula C9H6O3
MDL Number MFCD00005848
CAS 496-41-3
Beilstein 124204
Merck Index 14,2561
Synonym Coumarilic acid
Boiling Point 310° to 315°C
Melting Point 192° to 196°C
Health Hazard 1 Warning
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number DF6490000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.