missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzo[b]furan-2-carboxaldehyde, 97%, Maybridge™

Chemical Identifiers

CAS 4265-16-1
Molecular Formula C9H6O2
Molecular Weight (g/mol) 146.145
InChI Key ADDZHRRCUWNSCS-UHFFFAOYSA-N
Synonym 1-benzofuran-2-carboxaldehyde, 2-benzofurancarbaldehyde, 2-benzofurancarboxaldehyde, 2-formylbenzofuran, benzo b furan-2-carboxaldehyde, benzo b-2-furfural, benzofuran-2-aldehyde, benzofuran-2-carbaldehyde, benzofuran-2-carboxaldehyde, coumarilaldehyde
PubChem CID 61341
IUPAC Name 1-benzofuran-2-carbaldehyde
SMILES C1=CC=C2C(=C1)C=C(O2)C=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC06604EA
SDS View Documents
Fisher Scientific Company
CC06604EA
10g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

4265-16-1
146.145
1-benzofuran-2-carboxaldehyde, 2-benzofurancarbaldehyde, 2-benzofurancarboxaldehyde, 2-formylbenzofuran, benzo b furan-2-carboxaldehyde, benzo b-2-furfural, benzofuran-2-aldehyde, benzofuran-2-carbaldehyde, benzofuran-2-carboxaldehyde, coumarilaldehyde
1-benzofuran-2-carbaldehyde
C9H6O2
ADDZHRRCUWNSCS-UHFFFAOYSA-N
61341
C1=CC=C2C(=C1)C=C(O2)C=O

Specifications

10g
96.5
97%
1-benzofuran-2-carboxaldehyde, 2-benzofurancarbaldehyde, 2-benzofurancarboxaldehyde, 2-formylbenzofuran, benzo b furan-2-carboxaldehyde, benzo b-2-furfural, benzofuran-2-aldehyde, benzofuran-2-carbaldehyde, benzofuran-2-carboxaldehyde, coumarilaldehyde
C1=CC=C2C(=C1)C=C(O2)C=O
146.145
146.14
1-Benzofuran-2-carbaldehyde
4265-16-1
100.0
C9H6O2
ADDZHRRCUWNSCS-UHFFFAOYSA-N
1-benzofuran-2-carbaldehyde
61341
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.