missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzo[a]pyrene, 96%, Alfa Aesar™

Extensively used in cancer research

Manufacturer:  Alfa Aesar 15856MC

 View more versions of this product

Catalog No. AA15856MC



Specifications

Specifications

Benzo[a]pyrene
Undesignated
100mg
C20H12
UN3077
benzo a pyrene, 3,4-benzopyrene, benzo a pyrene, benzo pqr tetraphene, 3,4-benzpyrene, benzpyrene, benzo def chrysene, 6,7-benzopyrene, benz a pyrene, 3,4-benz a pyrene
FMMWHPNWAFZXNH-UHFFFAOYSA-N
benzo[a]pyrene
2336
252.3
96%
495°C
177°C to 180°C
50-32-8
MFCD00003602
14,1103
Soluble in benzene, toluene, and xylene; Sparingly soluble in alcohol, methanol; Insoluble in water
C1=CC=C2C3=C4C(=CC2=C1)C=CC5=C4C(=CC=C5)C=C3
252.316
CHEBI:29865
Crystalline

Chemical Identifiers

50-32-8
252.316
FMMWHPNWAFZXNH-UHFFFAOYSA-N
2336
benzo[a]pyrene
C20H12
MFCD00003602
benzo a pyrene, 3,4-benzopyrene, benzo a pyrene, benzo pqr tetraphene, 3,4-benzpyrene, benzpyrene, benzo def chrysene, 6,7-benzopyrene, benz a pyrene, 3,4-benz a pyrene
CHEBI:29865
C1=CC=C2C3=C4C(=CC2=C1)C=CC5=C4C(=CC=C5)C=C3
Safety and Handling

Safety and Handling

GHS H Statement
H340-H350-H360-H317
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : DJ3675000

TSCA : TSCA

SDS Documents