Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzimidazole-6-thiocarboxamide, 97%

Chemical Identifiers

Chemical Name or Material Benzimidazole-6-thiocarboxamide
Formula Weight 177.23
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6668003

 
alfa aesar™
H6668003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66680MD

 
alfa aesar™
H66680MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 177.23
Chemical Name or Material Benzimidazole-6-thiocarboxamide
Assay 97%
Physical Form Solid
Color Yellow to pale brown
Quantity 1g