Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzimidazole-6-carboxamidoxime, 97%

Chemical Identifiers

Chemical Name or Material Benzimidazole-6-carboxamidoxime
Formula Weight 176.18
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6640203

 
alfa aesar™
H6640203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66402MD

 
alfa aesar™
H66402MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 176.18
Chemical Name or Material Benzimidazole-6-carboxamidoxime
Assay 97%
Physical Form Solid
Quantity 1g
Color Pale brown