Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzimidazole-5-boronic acid pinacol ester, 95%

Chemical Identifiers

Chemical Name or Material Benzimidazole-5-boronic acid pinacol ester
CAS 1007206-54-3
Formula Weight 244.1
MDL Number MFCD11054041
Beilstein 20872316
Synonym 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzimidazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6698803

 
alfa aesar™
H6698803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66988MD

 
alfa aesar™
H66988MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 202° to 206°C
Formula Weight 244.1
Chemical Name or Material Benzimidazole-5-boronic acid pinacol ester
Synonym 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzimidazole
CAS 1007206-54-3
MDL Number MFCD11054041
Assay 95%
Quantity 1g
Beilstein 20872316
Physical Form Solid