Please login to your online account to display your discounted pricing

Benzilic Acid +99%, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  105425000

 View more versions of this product

Catalog No. AC105425000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Formula Weight 228.25
Physical Form Powder
Quantity 500g
Color White to cream-white
Beilstein 10, 342
Merck Index 15, 1082
EINECS Number 200-993-2
TSCA TSCA
Melting Point 148° to 152°C
RTECS Number DD2064000
CAS Min % 99.0
CAS Max % 100.0
Chemical Name or Material Benzilic acid
Infrared Spectrum Authentic
Name Note 99+%
Assay Percent Range 99+%
Molecular Formula C14H12O3
Linear Formula (C6H5)2C(OH)CO2H
MDL Number MFCD00004447
CAS 76-93-7
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Plastic bottle
Synonym Diphenylglycolic acid

Specifications

Chemical Name or Material Benzilic acid
Name Note 99+%
CAS 76-93-7
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C14H12O3
Linear Formula (C6H5)2C(OH)CO2H
Formula Weight 228.25
MDL Number MFCD00004447
Synonym Diphenylglycolic acid
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.

SDS & Safety Documents