Please login to your online account to display your discounted pricing

Benzilic Acid +99%, ACROS Organics™

$122.46 - $122.46

Chemical Identifiers

Chemical Name or Material Benzilic acid
Name Note 99+%
CAS 76-93-7
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C14H12O3
Linear Formula (C6H5)2C(OH)CO2H
Formula Weight 228.25
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC105425000

  SDS
thermo fisher scientific
105425000
Plastic bottle 500g Each for $122.46
Description & Specifications

Specifications

Assay ≥99.0%
Formula Weight 228.25
Physical Form Powder
Quantity 500g
Color White to cream-white
Merck Index 15, 1082
Beilstein 10, 342
TSCA TSCA
RTECS Number DD2064000
EINECS Number 200-993-2
Melting Point 148° to 152°C
CAS Min % 99.0
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Linear Formula (C6H5)2C(OH)CO2H
MDL Number MFCD00004447
Chemical Name or Material Benzilic acid
Name Note 99+%
CAS 76-93-7
Molecular Formula C14H12O3
Assay Percent Range 99+%
Synonym Diphenylglycolic acid
Packaging Plastic bottle
Safety and Handling

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.