missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzil monohydrazone, 98%, ACROS Organics™

$69.27 - $69.27

Chemical Identifiers

CAS 5344-88-7
Molecular Formula C14H12N2O
Molecular Weight (g/mol) 224.263
MDL Number MFCD00007625
InChI Key CDQPGWNBSOSEMZ-DTQAZKPQSA-N
Synonym 1, hydrazone, 2-hydrazinylidene-1,2-diphenylethan-1-one, 2-hydrazono-1,2-diphenyl-ethanone, 2-hydrazono-1,2-diphenylethanone, 2e-2-hydrazinylidene-1,2-diphenylethanone, 2z-2-hydrazinylidene-1,2-diphenylethanone, benzil monohydrazone, cdqpgwnbsosemz-dtqazkpqsa, ethanedione, diphenyl-, monohydrazone, hydrazonodeoxybenzoin
PubChem CID 5538956
IUPAC Name (2E)-2-hydrazinylidene-1,2-diphenylethanone
SMILES C1=CC=C(C=C1)C(=NN)C(=O)C2=CC=CC=C2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC105430250
SDS View Documents
Thermo Fisher Scientific
105430250
25g Glass Bottle
Each for $69.27
 
Specifications

Chemical Identifiers

5344-88-7
224.263
CDQPGWNBSOSEMZ-DTQAZKPQSA-N
5538956
C1=CC=C(C=C1)C(=NN)C(=O)C2=CC=CC=C2
C14H12N2O
MFCD00007625
1, hydrazone, 2-hydrazinylidene-1,2-diphenylethan-1-one, 2-hydrazono-1,2-diphenyl-ethanone, 2-hydrazono-1,2-diphenylethanone, 2e-2-hydrazinylidene-1,2-diphenylethanone, 2z-2-hydrazinylidene-1,2-diphenylethanone, benzil monohydrazone, cdqpgwnbsosemz-dtqazkpqsa, ethanedione, diphenyl-, monohydrazone, hydrazonodeoxybenzoin
(2E)-2-hydrazinylidene-1,2-diphenylethanone

Specifications

Authentic
Yellow
25g
97.5
98%
C6H5C(=NNH2)COC6H5
1, hydrazone, 2-hydrazinylidene-1,2-diphenylethan-1-one, 2-hydrazono-1,2-diphenyl-ethanone, 2-hydrazono-1,2-diphenylethanone, 2e-2-hydrazinylidene-1,2-diphenylethanone, 2z-2-hydrazinylidene-1,2-diphenylethanone, benzil monohydrazone, cdqpgwnbsosemz-dtqazkpqsa, ethanedione, diphenyl-, monohydrazone, hydrazonodeoxybenzoin
C1=CC=C(C=C1)C(=NN)C(=O)C2=CC=CC=C2
224.263
224.26
98%
Glass Bottle
147.0°C to 151.0°C
5344-88-7
100.0
C14H12N2O
MFCD00007625
CDQPGWNBSOSEMZ-DTQAZKPQSA-N
(2E)-2-hydrazinylidene-1,2-diphenylethanone
5538956
Very Fine Crystalline Powder
Benzil monohydrazone, 98%
Safety and Handling

Safety and Handling

EINECSNumber : 226-292-1

SDS