Please login to your online account to display your discounted pricing

Benzil monohydrazone, 98%, ACROS Organics™


Quantity 5g
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  105430050

 View more versions of this product

Catalog No. AC105430050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Very Fine Crystalline Powder
Assay ≥97.5%
Formula Weight 224.26
Quantity 5g
Color Very slightly yellow
EINECS Number 226-292-1
Melting Point 147° to 151°C
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass Bottle
Name Note 98%
Linear Formula C6H5C(=NNH2)COC6H5
MDL Number MFCD00007625
Chemical Name or Material Benzil monohydrazone
Assay Percent Range 98%
Molecular Formula C14H12N2O
CAS 5344-88-7
CAS Min % 97.5

Specifications

Chemical Name or Material Benzil monohydrazone
Name Note 98%
CAS 5344-88-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C14H12N2O
Linear Formula C6H5C(=NNH2)COC6H5
Formula Weight 224.26
MDL Number MFCD00007625