missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(Benzhydrylthio)acetic Acid, TCI America™

$152.78 - $459.88

Chemical Identifiers

CAS 63547-22-8
Molecular Formula C15H14O2S
Molecular Weight (g/mol) 258.335
MDL Number MFCD05262040
InChI Key HTHFEDOFDBZPRX-UHFFFAOYSA-N
Synonym (Diphenylmethylthio)acetic Acid
PubChem CID 2077886
IUPAC Name 2-benzhydrylsulfanylacetic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)SCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B41755G View Documents TCI America
B41755G
5g Each for $152.78
 
B417525G View Documents TCI America
B417525G
25g Each for $459.88
 
Specifications

Chemical Identifiers

63547-22-8
258.335
HTHFEDOFDBZPRX-UHFFFAOYSA-N
2077886
C1=CC=C(C=C1)C(C2=CC=CC=C2)SCC(=O)O
C15H14O2S
MFCD05262040
(Diphenylmethylthio)acetic Acid
2-benzhydrylsulfanylacetic acid

Specifications

White-Yellow
5g
C15H14O2S
(Diphenylmethylthio)acetic Acid
C1=CC=C(C=C1)C(C2=CC=CC=C2)SCC(=O)O
258.335
258.34
≥98.0% (GC,T)
130°C
63547-22-8
MFCD05262040
HTHFEDOFDBZPRX-UHFFFAOYSA-N
2-benzhydrylsulfanylacetic acid
2077886
Crystal-Powder at 20°C
(Benzhydrylthio)acetic Acid
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer