Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzhydrylidenefluorene, 98+%

$35.11 - $367.65

Chemical Identifiers

Chemical Name or Material Benzhydrylidenefluorene
CAS 4709-68-6
Molecular Formula C26H18
Formula Weight 330.43
MDL Number MFCD00051452
Beilstein 1977991
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2472203

  SDS
alfa aesar™
B2472203
1g Each for $35.11

AAB2472214

  SDS
alfa aesar™
B2472214
25g Each for $367.65

AAB2472206

  SDS
alfa aesar™
B2472206
5g Each for $83.93
Description & Specifications

Specifications

Formula Weight 330.43
Chemical Name or Material Benzhydrylidenefluorene
Assay ≥98%
Molecular Formula C26H18
MDL Number MFCD00051452
CAS 4709-68-6
Beilstein 1977991
Melting Point 230° to 232°C
Quantity 1g