missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzhydrylamine Hydrochloride 98.0+%, TCI America™
$108.11 - $501.43
Chemical Identifiers
| CAS | 5267-34-5 |
|---|---|
| Molecular Formula | C13H14ClN |
| Molecular Weight (g/mol) | 219.712 |
| MDL Number | MFCD00013023 |
| InChI Key | CIHWJRSPVJBHGT-UHFFFAOYSA-N |
| Synonym | benzhydrylamine hydrochloride, aminodiphenylmethane hydrochloride, benzhydrylamine, hydrochloride, diphenylmethanamine hydrochloride, benzhydrylammonium chloride, aminodiphenylmethane hcl, 1,1-diphenylmethylamine hydrochloride, diphenylmethylamine, chloride, benzhydrylamin hydrochloride, bha-polystyrene |
| PubChem CID | 78921 |
| IUPAC Name | diphenylmethanamine;hydrochloride |
| SMILES | C1=CC=C(C=C1)C(C2=CC=CC=C2)N.Cl |
Chemical Identifiers
| 5267-34-5 | |
| 219.712 | |
| CIHWJRSPVJBHGT-UHFFFAOYSA-N | |
| 78921 | |
| C1=CC=C(C=C1)C(C2=CC=CC=C2)N.Cl |
| C13H14ClN | |
| MFCD00013023 | |
| benzhydrylamine hydrochloride, aminodiphenylmethane hydrochloride, benzhydrylamine, hydrochloride, diphenylmethanamine hydrochloride, benzhydrylammonium chloride, aminodiphenylmethane hcl, 1,1-diphenylmethylamine hydrochloride, diphenylmethylamine, chloride, benzhydrylamin hydrochloride, bha-polystyrene | |
| diphenylmethanamine;hydrochloride |
Specifications
| 5267-34-5 | |
| White | |
| MFCD00013023 | |
| benzhydrylamine hydrochloride, aminodiphenylmethane hydrochloride, benzhydrylamine, hydrochloride, diphenylmethanamine hydrochloride, benzhydrylammonium chloride, aminodiphenylmethane hcl, 1,1-diphenylmethylamine hydrochloride, diphenylmethylamine, chloride, benzhydrylamin hydrochloride, bha-polystyrene | |
| C1=CC=C(C=C1)C(C2=CC=CC=C2)N.Cl | |
| 219.712 | |
| 219.71 | |
| Crystalline Powder |
| 293°C | |
| C13H14ClN | |
| 25 g | |
| CIHWJRSPVJBHGT-UHFFFAOYSA-N | |
| diphenylmethanamine;hydrochloride | |
| 78921 | |
| ≥98.0% (HPLC,T) | |
| Benzhydrylamine Hydrochloride |
Safety and Handling
TSCA : No