missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzhydrylamine hydrochloride, polymer-supported, 1% cross-linked, 100-200 mesh, 0.5-1.5 mmol/g on PS-DVB

Manufacturer:  Alfa Aesar™ 4445914

 View more versions of this product

Catalog No. AA4445914

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Benzhydrylamine hydrochloride
25g
5267-34-5
MFCD03456188
CIHWJRSPVJBHGT-UHFFFAOYSA-N
diphenylmethanamine;hydrochloride
78921
100 to 200 Mesh
Polymer-supported, 1% cross-linked, 0.5 to 1.5 mmol/g on PS-DVB
C13H14ClN
benzhydrylamine hydrochloride, aminodiphenylmethane hydrochloride, benzhydrylamine, hydrochloride, diphenylmethanamine hydrochloride, benzhydrylammonium chloride, aminodiphenylmethane hcl, 1,1-diphenylmethylamine hydrochloride, diphenylmethylamine, chloride, benzhydrylamin hydrochloride, bha-polystyrene
C1=CC=C(C=C1)C(C2=CC=CC=C2)N.Cl
219.712
Powder

Chemical Identifiers

5267-34-5
219.712
CIHWJRSPVJBHGT-UHFFFAOYSA-N
78921
C1=CC=C(C=C1)C(C2=CC=CC=C2)N.Cl
C13H14ClN
MFCD03456188
benzhydrylamine hydrochloride, aminodiphenylmethane hydrochloride, benzhydrylamine, hydrochloride, diphenylmethanamine hydrochloride, benzhydrylammonium chloride, aminodiphenylmethane hcl, 1,1-diphenylmethylamine hydrochloride, diphenylmethylamine, chloride, benzhydrylamin hydrochloride, bha-polystyrene
diphenylmethanamine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents