Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzhydrylamine hydrochloride, 97%

$33.91 - $130.64

Chemical Identifiers

Chemical Name or Material Benzhydrylamine hydrochloride
CAS 5267-34-5
Molecular Formula C13H13N·HCl
Formula Weight 219.72
MDL Number MFCD00013023
Beilstein 3914818
Merck Index 14,1076
Synonym Aminodiphenylmethane hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0238809

  SDS
alfa aesar™
L0238809
10g Each for $33.91

AAL0238818

  SDS
alfa aesar™
L0238818
50g Each for $130.64
Description & Specifications

Specifications

Formula Weight 219.72
Chemical Name or Material Benzhydrylamine hydrochloride
MDL Number MFCD00013023
Molecular Formula C13H13N·HCl
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 5267-34-5
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 10g
Melting Point ≈298°C
Beilstein 3914818
Merck Index 14,1076
Synonym Aminodiphenylmethane hydrochloride
Assay 97%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.