Learn More
Benzhydryl chloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2476436
Description
Benzhydryl chloride was used as initiator during the controlled radical polymerization of styrene catalyzed by ionic iron complex. It was also used as the starting reagent in the synthesis of trimethylhydroquinone derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzhydryl chloride was used as initiator during the controlled radical polymerization of styrene catalyzed by ionic iron complex. It was also used as the starting reagent in the synthesis of trimethylhydroquinone derivatives.
Solubility
Insoluble in water.
Notes
Store in a tightly sealed container. Store away from oxidizing agents, bases.
Specifications
Benzhydryl chloride | |
17°C to 20°C | |
139°C to 140°C (3 mmHg) | |
Pungent | |
1.595 | |
500 g | |
607925 | |
Insoluble in water. | |
C1=CC=C(C=C1)C(C2=CC=CC=C2)Cl | |
202.681 | |
202.69 |
90-99-3 | |
1.14 | |
160°C (320°F) | |
C13H11Cl | |
MFCD00000855 | |
UN3265 | |
benzhydryl chloride, chlorodiphenylmethane, chloromethylene dibenzene, diphenylchloromethane, diphenylmethyl chloride, benzene, 1,1'-chloromethylene bis, chloro phenyl methyl benzene, methane, chlorodiphenyl, unii-cn9n9ayv4b, 1,1'-chloromethylene bisbenzene | |
ZDVDCDLBOLSVGM-UHFFFAOYSA-N | |
[chloro(phenyl)methyl]benzene | |
7035 | |
98% |
Chemical Identifiers
90-99-3 | |
202.681 | |
ZDVDCDLBOLSVGM-UHFFFAOYSA-N | |
7035 | |
C1=CC=C(C=C1)C(C2=CC=CC=C2)Cl |
C13H11Cl | |
MFCD00000855 | |
benzhydryl chloride, chlorodiphenylmethane, chloromethylene dibenzene, diphenylchloromethane, diphenylmethyl chloride, benzene, 1,1'-chloromethylene bis, chloro phenyl methyl benzene, methane, chlorodiphenyl, unii-cn9n9ayv4b, 1,1'-chloromethylene bisbenzene | |
[chloro(phenyl)methyl]benzene |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P234-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 202-031-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only