missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzhydrazide, 98%

Manufacturer:  Alfa Aesar™ A1226822

 View more versions of this product

Catalog No. AAA1226822


 

Specifications

Specifications

Benzhydrazide
98%
MFCD00007596
Benzoic acid hydrazide; Benzoylhydrazine
C1=CC=C(C=C1)C(=O)NN
136.154
CHEBI:38454
113°C to 117°C
613-94-5
C7H8N2O
471797
WARCRYXKINZHGQ-UHFFFAOYSA-N
benzohydrazide
11955
136.15
100g

Chemical Identifiers

613-94-5
136.154
WARCRYXKINZHGQ-UHFFFAOYSA-N
11955
C7H8N2O
MFCD00007596
Benzoic acid hydrazide; Benzoylhydrazine
CHEBI:38454
Safety and Handling

Safety and Handling

GHS H Statement
H351
Suspected of causing cancer.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DH1575000

TSCA : TSCA

SDS Documents