missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzethonium chloride, 97%, Thermo Scientific™

Benzethonium chloride, 97%, C27H42ClNO2, CAS Number-121-54-0, benzethoniumchloride, benzethonium chloride, disilyn, hyamine, kylacol, phemeride, phemerol chloride, phemithyn, quatrachlor, solamin, 2.5kg, 96% min. (on dry substance) (Argentometry), 100.0, 96.0, CHEBI:31264, White, 204-479-9, 01,433

$82.77 - $270.94

Chemical Identifiers

CAS 121-54-0
Molecular Formula C27H42ClNO2
Molecular Weight (g/mol) 448.08
MDL Number MFCD00011742
InChI Key UREZNYTWGJKWBI-UHFFFAOYSA-M
Synonym benzethonium chloride, benzethoniumchloride, disilyn, hyamine, kylacol, phemeride, phemerol chloride, phemithyn, quatrachlor, solamin
PubChem CID 8478
ChEBI CHEBI:31264
IUPAC Name benzyl-dimethyl-[2-[2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy]ethyl]azanium;chloride
SMILES CC(C)(C)CC(C)(C)C1=CC=C(C=C1)OCCOCC[N+](C)(C)CC2=CC=CC=C2.[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC105381000
SDS View Documents Promotion Details
Fisher Scientific Acros
105381000
100g Plastic bottle
Each for $82.77
 
AC105385000
SDS View Documents Promotion Details
Thermo Fisher Scientific
105385000
500g Plastic bottle
Each for $270.94
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

121-54-0
448.08
UREZNYTWGJKWBI-UHFFFAOYSA-M
8478
benzyl-dimethyl-[2-[2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy]ethyl]azanium;chloride
C27H42ClNO2
MFCD00011742
benzethonium chloride, benzethoniumchloride, disilyn, hyamine, kylacol, phemeride, phemerol chloride, phemithyn, quatrachlor, solamin
CHEBI:31264
CC(C)(C)CC(C)(C)C1=CC=C(C=C1)OCCOCC[N+](C)(C)CC2=CC=CC=C2.[Cl-]

Specifications

121-54-0
100.0
158.0°C to 163.0°C
96% min. (on dry substance) (Argentometry)
Authentic
Plastic bottle
15, 1076
benzethonium chloride, benzethoniumchloride, disilyn, hyamine, kylacol, phemeride, phemerol chloride, phemithyn, quatrachlor, solamin
UREZNYTWGJKWBI-UHFFFAOYSA-M
benzyl-dimethyl-[2-[2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy]ethyl]azanium;chloride
8478
448.08
97%
96.0
White
5% max. (105°C, 4 hrs)
C27H42ClNO2
MFCD00011742
01,433
100g
Solubility in water: soluble. Other solubilities: soluble in alcohol,acetone and chloroform
CC(C)(C)CC(C)(C)C1=CC=C(C=C1)OCCOCC[N+](C)(C)CC2=CC=CC=C2.[Cl-]
448.08
CHEBI:31264
Amorphous or Crystalline Powder
Benzethonium chloride, 97%
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.

GHS P Statement
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes

GHS Signal Word: Danger

EINECSNumber : 204-479-9

RTECSNumber : BO7175000

TSCA : TSCA

SDS
Product Certifications

RUO – Research Use Only