Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzenesulfonyl hydrazide, polymer-supported, 1.8-2.2 mmol/g on polystyrene

Chemical Identifiers

Chemical Name or Material Benzenesulfonyl hydrazide
Name Note polymer-supported, 1.8 to 2.2mmol/g on polystyrene
MDL Number MFCD08064450
Synonym Sulfonyl hydrazide on polystyrene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1946606

  SDS
alfa aesar™
L1946606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1946609

  SDS
alfa aesar™
L1946609
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Benzenesulfonyl hydrazide
Name Note polymer-supported, 1.8 to 2.2mmol/g on polystyrene
MDL Number MFCD08064450
Synonym Sulfonyl hydrazide on polystyrene
Recommended Storage 2° to 8°C
Quantity 5g
Safety and Handling

2° to 8°C