Please login to your online account to display your discounted pricing

Benzenesulfonyl Hydrazide, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Benzenesulfonyl hydrazide
CAS 80-17-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H8N2O2S
Linear Formula C6H5SO2NHNH2
Formula Weight 172.2
MDL Number MFCD00007583
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC105301000

  SDS
thermo fisher scientific
105301000
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Powder
Formula Weight 172.2
Quantity 100g
Color White
Beilstein 11, 52
Fieser 07,13
Merck Index 15, 1074
Melting Point 100° to 104°C
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula C6H5SO2NHNH2
MDL Number MFCD00007583
Synonym Porofor BSH
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Flammable solid.
Heating may cause a fire.
CAS 80-17-1
Chemical Name or Material Benzenesulfonyl hydrazide
Molecular Formula C6H8N2O2S
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/other combustible materials.
Keep only in original container.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use CO2, dry chemical, or foam.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/other combustible materials.
Keep only in original container.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use CO2, dry chemical, or foam.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable solid.
Heating may cause a fire.