Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzenesulfonyl chloride, polymer-supported, 1.8-2.2 mmol/g on polystyrene

Chemical Identifiers

Chemical Name or Material Benzenesulfonyl chloride
Name Note polymer-supported, 1.8 to 2.2mmol/g on polystyrene
MDL Number MFCD00211665
Synonym Chlorosulfonated polystyrene; 4-Chlorosulfonyl polystyrene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1946506

  SDS
alfa aesar™
L1946506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1946514

  SDS
alfa aesar™
L1946514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Benzenesulfonyl chloride
Name Note polymer-supported, 1.8 to 2.2mmol/g on polystyrene
Quantity 5g
MDL Number MFCD00211665
Synonym Chlorosulfonated polystyrene; 4-Chlorosulfonyl polystyrene