Please login to your online account to display your discounted pricing

Benzenesulfonyl Chloride 99%, ACROS Organics™


Quantity 10mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  148480100

 View more versions of this product

Catalog No. AC148480100

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 176.62
Physical Form Liquid
Quantity 10mL
Color Clear
TSCA TSCA
EINECS Number 202-636-6
RTECS Number DB8750000
Fieser 01,46; 05,22; 14,27
CAS Max % 100.0
Packaging Glass bottle
Free Acid 0.05% max.
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Contact with water liberates toxic gas.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Benzenesulfonyl chloride
Linear Formula C6H5SO2Cl
CAS Min % 98.5
Molecular Formula C6H5ClO2S
Assay Percent Range 99%
MDL Number MFCD00007426
Name Note 99%
CAS 98-09-9
Refractive Index 1.5508 to 1.5528 (20°C, 589nm)
Health Hazard 1 Danger

Specifications

Chemical Name or Material Benzenesulfonyl chloride
Name Note 99%
CAS 98-09-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H5ClO2S
Linear Formula C6H5SO2Cl
Formula Weight 176.62
MDL Number MFCD00007426
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Contact with water liberates toxic gas.

SDS & Safety Documents