Learn More
Benzenesulfonyl bromide, 95%, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific AC441340010
Specifications
Benzenesulfonyl bromide | |
94.0 | |
Yellow to Brown | |
83°C to 84°C (0.5 Torr) | |
Authentic | |
Glass bottle | |
1g | |
C1=CC=C(C=C1)S(=O)(=O)Br | |
221.07 | |
221.07 | |
95% |
2297-65-6 | |
100.0 | |
19.5°C | |
94% min. (GC) | |
C6H5BrO2S | |
phenylsulfonyl bromide | |
CGWWQPZGKPHLBU-UHFFFAOYSA-N | |
benzenesulfonyl bromide | |
11138664 | |
Liquid |
Chemical Identifiers
2297-65-6 | |
221.07 | |
phenylsulfonyl bromide | |
benzenesulfonyl bromide |
C6H5BrO2S | |
CGWWQPZGKPHLBU-UHFFFAOYSA-N | |
11138664 | |
C1=CC=C(C=C1)S(=O)(=O)Br |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO – Research Use Only