missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzenesulfonyl bromide, 95%, ACROS Organics™

Chemical Identifiers

CAS 2297-65-6
Molecular Formula C6H5BrO2S
Molecular Weight (g/mol) 221.07
InChI Key CGWWQPZGKPHLBU-UHFFFAOYSA-N
Synonym phenylsulfonyl bromide
PubChem CID 11138664
IUPAC Name benzenesulfonyl bromide
SMILES C1=CC=C(C=C1)S(=O)(=O)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC441340010
SDS View Documents
Thermo Fisher Scientific
AC441340010
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC441340050
SDS View Documents
Thermo Fisher Scientific
AC441340050
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

2297-65-6
221.07
phenylsulfonyl bromide
benzenesulfonyl bromide
C6H5BrO2S
CGWWQPZGKPHLBU-UHFFFAOYSA-N
11138664
C1=CC=C(C=C1)S(=O)(=O)Br

Specifications

Authentic
83°C to 84°C (0.5 Torr)
19.5°C
2297-65-6
100.0
C6H5BrO2S
CGWWQPZGKPHLBU-UHFFFAOYSA-N
benzenesulfonyl bromide
11138664
Liquid
Benzenesulfonyl bromide
Glass bottle
Yellow to Brown
1g
94.0
94% min. (GC)
phenylsulfonyl bromide
C1=CC=C(C=C1)S(=O)(=O)Br
221.07
221.07
95%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger