missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzenesulfonic acid, 90%, tech., ACROS Organics™

Manufacturer:  Acros Organics AC159020250

 View more versions of this product

Catalog No. AC159020250



Specifications

Specifications

Benzenesulfonic acid
Authentic
Yellow to Brown
25g
88.0
90%
C6H5SO3H
benzenesulphonic acid, phenylsulfonic acid, besylic acid, benzene sulphonic acid, benzenemonosulfonic acid, benzene sulfonic acid, kyselina benzensulfonova, benzensulfonic acid, ccris 4595, kyselina benzensulfonova czech
C1=CC=C(C=C1)S(=O)(=O)O
158.171
CHEBI:64455
Damp Crystalline Solid or Fused Mass
Technical
>100°C
Glass bottle
45.0°C to 50.0°C
98-11-3,7664-93-9
100.0
C6H6O3S
MFCD00011689
SRSXLGNVWSONIS-UHFFFAOYSA-N
benzenesulfonic acid
7371
158.17
90%

Chemical Identifiers

98-11-3
158.171
SRSXLGNVWSONIS-UHFFFAOYSA-N
7371
benzenesulfonic acid
C6H6O3S
MFCD00011689
benzenesulphonic acid, phenylsulfonic acid, besylic acid, benzene sulphonic acid, benzenemonosulfonic acid, benzene sulfonic acid, kyselina benzensulfonova, benzensulfonic acid, ccris 4595, kyselina benzensulfonova czech
CHEBI:64455
C1=CC=C(C=C1)S(=O)(=O)O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents