missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzenesulfonic acid, 90%, tech., ACROS Organics™

Manufacturer:  Acros Organics 159020050

 View more versions of this product

Catalog No. AC159020050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Benzenesulfonic acid
88.0
Authentic
Technical Grade
90%
C6H5SO3H
SRSXLGNVWSONIS-UHFFFAOYSA-N
benzenesulfonic acid
7371
Yellow through Brown
45.0°C to 50.0°C
5g
98-11-3,7664-93-9
100.0
Glass bottle
90%
C6H6O3S
MFCD00011689
C1=CC=C(C=C1)S(=O)(=O)O
158.171
CHEBI:64455
158.17
Damp Crystalline Solid or Fused Mass
>100°C

Chemical Identifiers

98-11-3
158.171
SRSXLGNVWSONIS-UHFFFAOYSA-N
CHEBI:64455
C6H6O3S
MFCD00011689
7371
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents