Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzenesulfinic acid copper(II) salt hydrate, 97%

$130.84 - $521.81

Chemical Identifiers

Chemical Name or Material Benzenesulfinic acid copper(II) salt hydrate
CAS 64586-62-5
Molecular Formula C12H10CuO4S2·xH2O
Formula Weight 345.88 (Anhydrous)
MDL Number MFCD00061578
Synonym Copper(II) benzenesulfinate hydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1661406

 
alfa aesar™
A1661406
5g Each for $130.84

AAA1661414

 
alfa aesar™
A1661414
25g Each for $521.81
Description & Specifications

Specifications

Formula Weight 345.88 (Anhydrous)
Molecular Formula C12H10CuO4S2·xH2O
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material Benzenesulfinic acid copper(II) salt hydrate
Assay 97%
MDL Number MFCD00061578
CAS 64586-62-5
Quantity 5g
Synonym Copper(II) benzenesulfinate hydrate
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.