Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzeneboronic acid, polymer-supported, 2.6-3.2 mmol/g

Chemical Identifiers

Chemical Name or Material Benzeneboronic Acid, Polymer-supported
MDL Number MFCD03456184
Synonym 4-Boronopolystyrene; Phenylboronic Acid, Polymer-supported
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1945903

 
alfa aesar™
L19459-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1945906

 
alfa aesar™
L19459-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1945914

 
alfa aesar™
L19459-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Concentration 2.6 to 3.2mmol/g
Quantity 1g
Chemical Name or Material Benzeneboronic acid, polymer-supported
MDL Number MFCD03456184
Synonym 4-Boronopolystyrene; Phenylboronic acid, polymer-supported