Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzene-1,3-dithiocarboxamide, 97%

Chemical Identifiers

Chemical Name or Material Benzene-1,3-dithiocarboxamide
Molecular Formula C8H8N2S2
Formula Weight 196.28
MDL Number MFCD16293899
UN Number UN2811
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5185303

 
alfa aesar™
H5185303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 196.28
Chemical Name or Material Benzene-1,3-dithiocarboxamide
Assay 97%
Molecular Formula C8H8N2S2
MDL Number MFCD16293899
Quantity 1g
UN Number UN2811
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P280e-P309-P310
Wear protective clothing and face protection.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280e-P309-P310
Wear protective clothing and face protection.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301
Toxic if swallowed.